2-(Methylthio)pyrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009618-5G 009618-1G 009618-25G
FLUO009618-5GEA 0 GBP
FLUO009618-5G FLUO009618-1G FLUO009618-25G
2-(Methylthio)pyrazine
2-(Methylthio)pyrazine
Formula: C₅H₆N₂S
MDL Number: MFCD00041024
CAS Number: 21948-70-9
EINECS: 244-675-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR