Iron(III) acetylacetonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')
M-Clarity™ quality level = MQ100
517003-10G
517003-50G
SIAL517003-10GEA
67.9
GBP
SIAL517003-10G
SIAL517003-50G
Iron(III) acetylacetonate, Sigma-Aldrich®
Iron (III) acetylacetonate
Formula:
C₁₅H₂₁FeO₆ MW: 353.18 g/mol Boiling Pt: 100 °C (9.7513 mmHg) Melting Pt: 180…183 °C Density: 5.24 g/cm³ (20 °C) |
MDL Number:
MFCD00000020 CAS Number: 14024-18-1 EINECS: 237-853-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...