Palladium(II) acetylacetonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Palladium (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')palladium (II), Palladium (II) bis(2,4-pentanedionato-O,O'), Palladium(II) 2,4-pentanedionate
M-Clarity™ quality level = MQ100
209015-1G
209015-5G
SIAL209015-1GEA
113
GBP
SIAL209015-1G
SIAL209015-5G
Palladium(II) acetylacetonate, Sigma-Aldrich®
Palladium (II) acetylacetonate
Formula:
C₁₀H₁₄O₄Pd Boiling Pt: ∼190 °C (1013 hPa) Melting Pt: 190 °C |
MDL Number:
MFCD00000025 CAS Number: 14024-61-4 EINECS: 237-859-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...