Zirconium(IV) tert-butanolate ≥99% (metals basis)
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Zirconium(IV) t-butoxide, Zirconium (IV) tert-butoxide, Zirconium (IV) tert-butylate
39547.03
39547.06
39547.14
39547.03EA
40
GBP
39547.03
39547.06
39547.14
Zirconium(IV) tert-butanolate ≥99% (metals basis)
Zirconium (IV) tert-butanolate
Purity: 99%
CAS No.: 2081-12-1
Molecular Formula: C16H36O4Zr
Formula Weight: 383.68
Flash Point: 85°(185°F)
Physical Form: Liquid in ampoules
Density: 0.985
Sensitivity: Moisture and Light Sensitive
Appearance: Yellow
Boiling Point: 90°/5mm
Beilstein Reference: 3681870
MDL No.: MFCD00075085
Formula:
C₁₆H₃₆O₄Zr MW: 383.68 g/mol Boiling Pt: 90 °C (5 mmHg) Density: 0.985 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075085 CAS Number: 2081-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...