Silver hexafluoroantimonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Silver hexafluoroantimonate(V)

M-Clarity™ quality level = MQ100

227730-25G 227730-1G 227730-5G
SIAL227730-25GEA 532 GBP
SIAL227730-25G SIAL227730-1G SIAL227730-5G
Silver hexafluoroantimonate, Sigma-Aldrich®
Silver hexafluoroantimonate
Formula: AgF₆Sb
MW: 343.62 g/mol
Density: 4.65…4.85 g/cm³ (20 °C)
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR