Silver hexafluoroantimonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

007268-5G 007268-25G 007268-100G
FLUO007268-5GEA 0 GBP
FLUO007268-5G FLUO007268-25G FLUO007268-100G
Silver hexafluoroantimonate
Silver hexafluoroantimonate
Formula: AgF₆Sb
Density: 4.65…4.85 g/cm³ (20 °C)
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR