Silver(I) hexafluorophosphate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Silver hexafluorophosphate

002864-1G 002864-10G 002864-50G
FLUO002864-1GEA 0 GBP
FLUO002864-1G FLUO002864-10G FLUO002864-50G
Silver(I) hexafluorophosphate
Silver (I) hexafluorophosphate
Formula: AgF₆P
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR