Silver tetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Argentous tetrafluoroborate
M-Clarity™ quality level = MQ100
483052-25G
483052-5G
SIAL483052-25GEA
843
GBP
SIAL483052-25G
SIAL483052-5G
Silver tetrafluoroborate, Sigma-Aldrich®
Silver tetrafluoroborate
Formula:
AgBF₄ MW: 194.67 g/mol Melting Pt: 200 °C |
MDL Number:
MFCD00003408 CAS Number: 14104-20-2 EINECS: 237-956-5 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...