Silver tetrafluoroborate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Argentous tetrafluoroborate

M-Clarity™ quality level = MQ200

208361-10G 208361-1G 208361-50G
SIAL208361-10GEA 162 GBP
SIAL208361-10G SIAL208361-1G SIAL208361-50G
Silver tetrafluoroborate, Sigma-Aldrich®
Silver tetrafluoroborate
Formula: AgBF₄
MW: 194.67 g/mol
Melting Pt: 200
MDL Number: MFCD00003408
CAS Number: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR