Ammonium hexafluorophosphate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003135-100G 003135-500G 003135-25G
FLUO003135-100GEA 0 GBP
FLUO003135-100G FLUO003135-500G FLUO003135-25G
Ammonium hexafluorophosphate
Ammonium hexafluorophosphate
Formula: H₄F₆NP
MW: 163 g/mol
Melting Pt: 198
Density: 2.18 g/cm³ (18 °C)
MDL Number: MFCD00064642
CAS Number: 16941-11-0
EINECS: 241-009-1
UN: 3260
ADR: 8 III
Merck Index: 12,00557

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR