Gallium(III) acetylacetonate ≥99.99%, PURATREM

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Tris(2,4-pentanedionato-O,O')gallium (III), Gallium (III) 2,4-pentanedionate, Gallium (III) tris(2,4-pentanedionato-O,O'), Gallium(III) Acetylacetonate, Acetylacetone Gallium(III) salt, Tris(2,4-pentanedionato)gallium(III), Ga(acac)3

93-3130-25G AB120512.0025 93-3130-5G
STRE93-3130-25GEA 233.53 GBP
STRE93-3130-25G ABCRAB120512.0025 STRE93-3130-5G
Gallium(III) acetylacetonate ≥99.99%, PURATREM
Gallium (III) acetylacetonate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₅H₂₁GaO₆
Boiling Pt: 140 °C (10 mmHg) (subl.)
Melting Pt: 192…194 °CC >280 °C (dec.)
Density: 1.42
MDL Number: MFCD00013492
CAS Number: 14405-43-7
EINECS: 238-377-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR