Barium titanate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
M-Clarity™ quality level = MQ100
467634-100G
467634-25G
SIAL467634-100GEA
470
GBP
SIAL467634-100G
SIAL467634-25G
Barium titanate, Sigma-Aldrich®
Barium titanate
Formula:
BaO₃Ti Melting Pt: 1625 °C Density: 5.67 g/cm³ (20 °C) |
MDL Number:
MFCD00003447 CAS Number: 12047-27-7 EINECS: 234-975-0 UN: 1564 ADR: 6.1,III Merck Index: 12,01029 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...