Barium titanate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

M-Clarity™ quality level = MQ100

208108-2KG 208108-500G
SIAL208108-2KGEA 290 GBP
SIAL208108-2KG SIAL208108-500G
Barium titanate, Sigma-Aldrich®
Barium titanate
Formula: BaO₃Ti
MW: 233.21 g/mol
Melting Pt: 1625 °C
Density: 5.67 g/cm³ (20 °C)
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01029

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR