Tungstic(VI) acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dihydrogen wolframate, Tungstic acid

M-Clarity™ quality level = MQ100

223328-100G 223328-500G
SIAL223328-100GEA 94.5 GBP
SIAL223328-100G SIAL223328-500G
Tungstic(VI) acid, Sigma-Aldrich®
Tungstic (VI) acid
Formula: H₂O₄W
MW: 249.85 g/mol
MDL Number: MFCD00011336
CAS Number: 7783-03-1
EINECS: 231-975-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR