4-Aminoantipyrine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone
M-Clarity™ quality level = MQ200
33528-100G-R
33528-25G-R
SIAL33528-100G-REA
109
GBP
SIAL33528-100G-R
SIAL33528-25G-R
4-Aminoantipyrine, Sigma-Aldrich®
4-Aminoantipyrine
Formula:
C₁₁H₁₃N₃O Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003145 CAS Number: 83-07-8 EINECS: 201-452-3 Merck Index: 13,00596 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...