1-(3,5-Xylyl)piperazin
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
019231-5G
019231-1G
FLUO019231-5GEA
0
GBP
FLUO019231-5G
FLUO019231-1G
1-(3,5-Xylyl)piperazin
1-(3,5-Xylyl)piperazin
Formula:
C₁₂H₁₈N₂ MW: 190.29 g/mol |
MDL Number:
MFCD03095639 CAS Number: 105907-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...