1-(3,4-Xylyl)piperazin
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(3,4-Xylyl)piperazine, 1-(3,4-Dimethylphenyl)piperazine
009779-5G
009779-25G
FLUO009779-5GEA
0
GBP
FLUO009779-5G
FLUO009779-25G
1-(3,4-Xylyl)piperazin
1-(3,4-Xylyl)piperazin
Formula:
C₁₂H₁₈N₂ MW: 190.29 g/mol Boiling Pt: 135 °C (0.6 mmHg) Melting Pt: 62 °C |
MDL Number:
MFCD00023129 CAS Number: 1014-05-7 EINECS: 213-798-2 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...