1-(2,6-Xylyl)piperazin

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021549-25G 021549-5G
FLUO021549-25GEA 0 GBP
FLUO021549-25G FLUO021549-5G
1-(2,6-Xylyl)piperazin
1-(2,6-Xylyl)piperazin
Formula: C₁₂H₁₈N₂
Boiling Pt: 148 °C (10 mmHg)
MDL Number: MFCD00038009
CAS Number: 1012-91-5
EINECS: 213-791-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR