1-(2,3-Xylyl)piperazin

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,3-Xylyl)piperazine, 1-(2,3-Dimethylphenyl)piperazine

009778-1G 009778-5G 009778-25G
FLUO009778-1GEA 0 GBP
FLUO009778-1G FLUO009778-5G FLUO009778-25G
1-(2,3-Xylyl)piperazin
1-(2,3-Xylyl)piperazin
Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Boiling Pt: 94 °C (0.3 mmHg)
MDL Number: MFCD00040730
CAS Number: 1013-22-5
EINECS: 213-794-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR