1-(4-Nitrophenyl)piperazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009817-5G
009817-25G
FLUO009817-5GEA
0
GBP
FLUO009817-5G
FLUO009817-25G
1-(4-Nitrophenyl)piperazine
1-(4-Nitrophenyl)piperazine
Formula:
C₁₀H₁₃N₃O₂ Melting Pt: 136.85 °C |
MDL Number:
MFCD00005961 CAS Number: 6269-89-2 EINECS: 228-443-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...