1-(2-Nitrophenyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019232-1G 019232-25G
FLUO019232-1GEA 0 GBP
FLUO019232-1G FLUO019232-25G
1-(2-Nitrophenyl)piperazine
1-(2-Nitrophenyl)piperazine
Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD00040728
CAS Number: 59084-06-9
EINECS: 261-593-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR