PIPES dipotassium salt, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid) dipotassium salt, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid dipotassium salt, PIPES-K2, Dipotassium piperazine-1,4-diethanesulphonate, Piperazine-N,N'-bis-2-ethanesulphonic acid dipotassium salt

M-Clarity™ quality level = MQ200

P7643-100G P7643-1KG P7643-25G P7643-500G
SIALP7643-100GEA 259 GBP
SIALP7643-100G SIALP7643-1KG SIALP7643-25G SIALP7643-500G
PIPES dipotassium salt, Sigma-Aldrich®
PIPES dipotassium salt
Formula: C₈H₁₆N₂O₆S₂K₂
MW: 378.55 g/mol
CAS Number: 108321-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR