1-(2-Hydroxyethyl)piperazine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: N-(2-Hydroxyethyl)piperazine, 2-(1-Piperazinyl)ethanol, 2-Piperazin-1-ylethanol

M-Clarity™ quality level = MQ200

H28807-100G H28807-500G
SIALH28807-100GEA 40.9 GBP
SIALH28807-100G SIALH28807-500G
1-(2-Hydroxyethyl)piperazine, Sigma-Aldrich®
1-(2-Hydroxyethyl)piperazine
Formula: C₆H₁₄N₂O
Boiling Pt: ∼246 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.0595 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005970
CAS Number: 103-76-4
EINECS: 203-142-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR