1-(2-Hydroxyethyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

005136-500G 005136-100G
FLUO005136-500GEA 0 GBP
FLUO005136-500G FLUO005136-100G
1-(2-Hydroxyethyl)piperazine
1-(2-Hydroxyethyl)piperazine
Formula: C₆H₁₄N₂O
Boiling Pt: ∼246 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1,0595 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005970
CAS Number: 103-76-4
EINECS: 203-142-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR