Homopiperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,4-Diazepane, 1,4-Diazacycloheptane, Perhydro-1,4-diazepine

021581-100G 021581-25G
FLUO021581-100GEA 0 GBP
FLUO021581-100G FLUO021581-25G
Homopiperazine
Homopiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 169 °C (1013 hPa)
Melting Pt: 43 °C
MDL Number: MFCD00006933
CAS Number: 505-66-8
EINECS: 208-016-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR