Isophorone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ300
W355305-1KG
W355305-20KG
W355305-5KG
W355305-SAMPLE
SIALW355305-1KGEA
69.2
GBP
SIALW355305-1KG
SIALW355305-20KG
SIALW355305-5KG
SIALW355305-SAMPLE
Isophorone, Sigma-Aldrich®
Isophorone
Formula:
C₉H₁₄O Boiling Pt: 215 °C (1013 hPa) Melting Pt: –8 °C Density: 0.922 g/cm³ (20 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001584 CAS Number: 78-59-1 EINECS: 201-126-0 Merck Index: 13,05215 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...