Isophorone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
I18709-100ML
I18709-1L
I18709-5ML
SIALI18709-100MLEA
25.8
GBP
SIALI18709-100ML
SIALI18709-1L
SIALI18709-5ML
Isophorone, Sigma-Aldrich®
Isophorone
Formula:
C₉H₁₄O Boiling Pt: 215 °C (1013 hPa) Melting Pt: –8 °C Density: 0.922 g/cm³ (20 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001584 CAS Number: 78-59-1 EINECS: 201-126-0 Merck Index: 13,05215 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...