1-Acetylcyclohexene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Acetyl-1-cyclohexene, Cyclohex-1-enylmethylketone, 1-(Cyclohex-1-en-1-yl)ethanone

M-Clarity™ quality level = MQ100

A14405-25G A14405-5G
SIALA14405-25GEA 209 GBP
SIALA14405-25G SIALA14405-5G
1-Acetylcyclohexene, Sigma-Aldrich®
1-Acetylcyclohexene
Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 202 °C (1013 hPa)
Density: 0.9704 g/cm³ (16 °C)
MDL Number: MFCD00001547
CAS Number: 932-66-1
EINECS: 213-256-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR