Cyclopropanecarbonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Cyclopropyl cyanide

005110-50G 005110-250G
FLUO005110-50GEA 0 GBP
FLUO005110-50G FLUO005110-250G
Cyclopropanecarbonitrile
Cyclopropanecarbonitrile
Formula: C₄H₅N
Boiling Pt: 133 °C (1013 hPa)
Melting Pt: –25 °C
Density: 0.911 g/cm³ (20 °C)
MDL Number: MFCD00001269
CAS Number: 5500-21-0
EINECS: 226-836-8
UN: 3275
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR