α,α-Dimethoxytoluene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Benzaldehyde dimethyl acetal

M-Clarity™ quality level = MQ300

W212806-1KG-K W212806-5KG-K
SIALW212806-1KG-KEA 195 GBP
SIALW212806-1KG-K SIALW212806-5KG-K
α,α-Dimethoxytoluene, Sigma-Aldrich®
α,α-Dimethoxytoluene
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 198 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
MDL Number: MFCD00008491
CAS Number: 1125-88-8
EINECS: 214-413-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR