4-Iodo-o-toluidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005444-5G
005444-25G
FLUO005444-5GEA
0
GBP
FLUO005444-5G
FLUO005444-25G
4-Iodo-o-toluidine
4-Iodo-o-toluidine
Formula:
C₇H₈IN Boiling Pt: 279 °C (1013 hPa) Melting Pt: 91…92 °C |
MDL Number:
MFCD00025299 CAS Number: 13194-68-8 EINECS: 236-154-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...