5-Iodo-o-toluidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-4-iodotoluene, 5-Iodo-2-methylaniline

M-Clarity™ quality level = MQ100

648647-1G 648647-5G
SIAL648647-1GEA 74.2 GBP
SIAL648647-1G SIAL648647-5G
5-Iodo-o-toluidine, Sigma-Aldrich®
5-Iodo-o-toluidine
Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 48…49 °C
MDL Number: MFCD00051531
CAS Number: 83863-33-6
EINECS: 281-094-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR