3-Iodo-p-toluidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
631264-25G
631264-5G
SIAL631264-25GEA
148
GBP
SIAL631264-25G
SIAL631264-5G
3-Iodo-p-toluidine, Sigma-Aldrich®
3-Iodo-p-toluidine
Formula:
C₇H₈IN MW: 233.05 g/mol |
MDL Number:
MFCD00047843 CAS Number: 35944-64-0 EINECS: 252-803-2 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...