2-Fluorobenzyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006202-5G 006202-1G
FLUO006202-5GEA 0 GBP
FLUO006202-5G FLUO006202-1G
2-Fluorobenzyl isothiocyanate
2-Fluorobenzyl isothiocyanate
Formula: C₈H₆FNS
MW: 167.21 g/mol
MDL Number: MFCD00046817
CAS Number: 64382-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR