2'-Fluoro-4'-hydroxyacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2-Fluoro-4-hydroxyphenyl)ethanone

009204-1G 009204-5G
FLUO009204-1GEA 0 GBP
FLUO009204-1G FLUO009204-5G
2'-Fluoro-4'-hydroxyacetophenone
2'-Fluoro-4'-hydroxyacetophenone
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00142713
CAS Number: 98619-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR