4-Chloro-3-iodobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004484-25G 004484-100G
FLUO004484-25GEA 0 GBP
FLUO004484-25G FLUO004484-100G
4-Chloro-3-iodobenzotrifluoride
4-Chloro-3-iodobenzotrifluoride
Formula: C₇H₃ClF₃I
Boiling Pt: 58 °C (1013 hPa)
Density: 1,92 g/cm³ (20 °C)
MDL Number: MFCD00001037
CAS Number: 672-57-1
EINECS: 211-593-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR