3'-Bromo-4'-fluoroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010205-10G
010205-100G
010205-25G
FLUO010205-10GEA
0
GBP
FLUO010205-10G
FLUO010205-100G
FLUO010205-25G
3'-Bromo-4'-fluoroacetophenone
3'-Bromo-4'-fluoroacetophenone
Formula:
C₈H₆BrFO Boiling Pt: 240 °C (1013 hPa) Melting Pt: 57 °C |
MDL Number:
MFCD00042466 CAS Number: 1007-15-4 EINECS: 213-750-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...