1,3-Dinitro-4-(trifluoromethoxy)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Dinitro-1-(trifluoromethoxy)benzene, 2,4-Dinitro(trifluoromethoxy)benzene

004970-25G 004970-100G 004970-250G
FLUO004970-25GEA 0 GBP
FLUO004970-25G FLUO004970-100G FLUO004970-250G
1,3-Dinitro-4-(trifluoromethoxy)benzene
1,3-Dinitro-4-(trifluoromethoxy)benzene
Formula: C₇H₃F₃N₂O₅
MW: 252.11 g/mol
MDL Number: MFCD00042268
CAS Number: 655-07-2
EINECS: 470-480-8
UN: 2927
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR