4-Fluoro-2-nitrophenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Fluoro-4-hydroxy-3-nitrobenzene
M-Clarity™ quality level = MQ100
345059-25G
345059-5G
SIAL345059-25GEA
77.5
GBP
SIAL345059-25G
SIAL345059-5G
4-Fluoro-2-nitrophenol, Sigma-Aldrich®
4-Fluoro-2-nitrophenol
Formula:
C₆H₄FNO₃ MW: 157.1 g/mol Melting Pt: 74 °C |
MDL Number:
MFCD00042528 CAS Number: 394-33-2 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...