4-Fluoro-2-nitrophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Fluoro-4-hydroxy-3-nitrobenzene

002388-5G 002388-25G 002388-100G
FLUO002388-5GEA 0 GBP
FLUO002388-5G FLUO002388-25G FLUO002388-100G
4-Fluoro-2-nitrophenol
4-Fluoro-2-nitrophenol
Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Melting Pt: 74 °C
MDL Number: MFCD00042528
CAS Number: 394-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR