4-Fluoro-2-iodoaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019309-5G 019309-25G
FLUO019309-5GEA 0 GBP
FLUO019309-5G FLUO019309-25G
4-Fluoro-2-iodoaniline
4-Fluoro-2-iodoaniline
Formula: C₆H₅FIN
MDL Number: MFCD00042584
CAS Number: 61272-76-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR