Propyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

P3130-100G P3130-500G
SIALP3130-100GEA 95.2 GBP
SIALP3130-100G SIALP3130-500G
Propyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Propyl 3,4,5-trihydroxybenzoate
Formula: C₁₀H₁₂O₅
Boiling Pt: 449 °C (1013 hPa)
Melting Pt: 145…148 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002196
CAS Number: 121-79-9
EINECS: 204-498-2
Merck Index: 12,08044

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR