Ethyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48640-100G-F
48640-500G-F
SIAL48640-100G-FEA
134
GBP
SIAL48640-100G-F
SIAL48640-500G-F
Ethyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Ethyl 3,4,5-trihydroxybenzoate
Formula:
C₉H₁₀O₅ |
MDL Number:
MFCD00016430 CAS Number: 831-61-8 EINECS: 212-608-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...