2-Bromo-6-chloro-4-fluoroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013190-5G
013190-100G
FLUO013190-5GEA
0
GBP
FLUO013190-5G
FLUO013190-100G
2-Bromo-6-chloro-4-fluoroaniline
2-Bromo-6-chloro-4-fluoroaniline
Formula:
C₆H₄BrClFN MW: 224.46 g/mol Boiling Pt: 260 °C (1013 hPa) Melting Pt: 55…56 °C |
MDL Number:
MFCD00051707 CAS Number: 201849-14-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...