Ethyl cinnamate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
NSC 6773, 3-Phenyl-acrylicacidethylester, Ethyl 3-phenyl-2-propenoate, Cinnamic acid ethyl ester, 3-Phenyl-2-propenoic acid ethyl ester
M-Clarity™ quality level = MQ300
W243000-10KG-K
W243000-1KG-K
W243000-25KG-K
SIALW243000-10KG-KEA
825
GBP
SIALW243000-10KG-K
SIALW243000-1KG-K
SIALW243000-25KG-K
Ethyl cinnamate, Sigma-Aldrich®
Ethyl cinnamate
Formula:
C₁₁H₁₂O₂ MW: 176.21 g/mol Boiling Pt: 271 Melting Pt: 7 Density: 1.046 g/cm³ (20 °C) |
MDL Number:
MFCD00009189 CAS Number: 103-36-6 EINECS: 203-104-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...