Dibutyl maleate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Maleic acid dibutyl ester

M-Clarity™ quality level = MQ200

D47102-1L D47102-250ML
SIALD47102-1LEA 111 GBP
SIALD47102-1L SIALD47102-250ML
Dibutyl maleate, Sigma-Aldrich®
Dibutyl maleate
Formula: C₁₂H₂₀O₄
Boiling Pt: 280 °C (1013 hPa)
Density: 0.9938 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009447
CAS Number: 105-76-0
EINECS: 203-328-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR