2-Phenylethyl acetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Phenylethyl Acetate, 2-Phenethyl acetate, Phenethyl acetate

M-Clarity™ quality level = MQ300

W285706-10KG-K W285706-1KG-K W285706-5KG-K
SIALW285706-10KG-KEA 318 GBP
SIALW285706-10KG-K SIALW285706-1KG-K SIALW285706-5KG-K
2-Phenylethyl acetate, Sigma-Aldrich®
2-Phenylethyl acetate
Formula: C₁₀H₁₂O₂
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.0286 g/cm³ (20 °C)
MDL Number: MFCD00008720
CAS Number: 103-45-7
EINECS: 203-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR