Ethyl isobutyrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methylpropionic acid ethyl ester
M-Clarity™ quality level = MQ100
246085-100ML
246085-500ML
SIAL246085-100MLEA
41.5
GBP
SIAL246085-100ML
SIAL246085-500ML
Ethyl isobutyrate, Sigma-Aldrich®
Ethyl isobutyrate
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 112 °C (1013 hPa) Melting Pt: –88 °C Density: 0.87 g/cm³ (20 °C) |
MDL Number:
MFCD00009165 CAS Number: 97-62-1 EINECS: 202-595-4 UN: 2385 ADR: 3,II Merck Index: 13,03847 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...