2-Propanesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Isopropylsulphonyl chloride

001772-5G 001772-25G 001772-100G
FLUO001772-5GEA 0 GBP
FLUO001772-5G FLUO001772-25G FLUO001772-100G
2-Propanesulphonyl chloride
2-Propanesulphonyl chloride
Formula: C₃H₇ClO₂S
MW: 142.61 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: –47 °C
Density: 1.273 g/cm³ (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00007453
CAS Number: 10147-37-2
EINECS: 233-415-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR