Butyl nitrite, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
226637-100G
226637-25G
SIAL226637-100GEA
256
GBP
SIAL226637-100G
SIAL226637-25G
Butyl nitrite, Sigma-Aldrich®
Butyl nitrite
Formula:
C₄H₉NO₂ Boiling Pt: 75 °C (1013 hPa) Density: 0.88 g/cm³ (20 °C) Flash Pt: 10 °C |
MDL Number:
MFCD00002058 CAS Number: 544-16-1 EINECS: 208-862-1 UN: 2351 ADR: 3,II Merck Index: 12,01617 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...